Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-1-Phenylbutylamine, ChiPros™, 98%, ee 98+%, Thermo Scientific™

Catalog No. AAH2704803
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2704803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2704803 Supplier Thermo Scientific Chemicals Supplier No. H2704803

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3789-60-4
Molecular Formula C10H15N
Molecular Weight (g/mol) 149.237
MDL Number MFCD08064291
InChI Key XHOXKVFLASIOJD-JTQLQIEISA-N
Synonym s-1-phenylbutylamine, s-1-phenylbutan-1-amine, 1s-1-phenylbutan-1-amine, pubchem24143, s-alpha-phenylbutylamine, s-1-phenyl-butylamine, s-1-phenyl-1-butanamine, s-1-phenylbutylamine, chipros, benzenemethanamine, a-propyl-, as, s-1-phenylbutylamine, chipros , ee 98+%
PubChem CID 2449444
IUPAC Name (1S)-1-phenylbutan-1-amine
SMILES CCCC(C1=CC=CC=C1)N

Specifications

Boiling Point 220°C
Quantity 1 g
UN Number UN2735
Sensitivity Air Sensitive
Formula Weight 149.24
Percent Purity 98%
Assay ee ≥98%
Chemical Name or Material (S)-1-Phenylbutylamine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.