Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-1-Phenylethylamine, 99+%, produced by BASF AG

Catalog No. p-3736914
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC371450500 50 g Glass bottle
AC371452500 250 g Glass bottle
AC371450010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC371450500 Supplier Thermo Scientific Chemicals Supplier No. 371450500
Only null left
Add to Cart

CAS: 2627-86-3 | C8H11N | 121.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chiral derivatization reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2627-86-3
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.18
MDL Number MFCD00064406
InChI Key RQEUFEKYXDPUSK-ZETCQYMHSA-N
Synonym s---1-phenylethylamine, s-1-phenylethanamine, 1s-1-phenylethanamine, s---alpha-methylbenzylamine, s-1-phenylethylamine, l--alpha-methylbenzylamine, l-alpha-methylbenzylamine, s-alpha-methylbenzenemethanamine, --alpha-phenethylamine, l---1-phenylethylamine
PubChem CID 75818
ChEBI CHEBI:35321
IUPAC Name (1S)-1-phenylethanamine
SMILES CC(C1=CC=CC=C1)N

Specifications

Melting Point -10.0°C
Color Colorless
Density 0.9400g/mL
Boiling Point 187.0°C to 189.0°C
Flash Point 71°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.5250 to 1.5270
Linear Formula C6H5CH(CH3)NH2
Quantity 50 g
Beilstein 12,II,587
Fieser 09,363; 13,185; 17,276; 12,319; 11,411; 05,441; 03,199; 01,838; 02,271; 08,393
Specific Gravity 0.94
Merck Index 13, 6054
Specific Rotation Condition −40.00 (20.00°C neat)
Specific Rotation − 40.00
Solubility Information Solubility in water: 42g/L (20°C). Other solubilities: soluble in most organic solvents,miscible with alcohols and ethers
Formula Weight 121.18
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material (S)-(-)-1-Phenylethylamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
EINECSNumber 220-098-

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.