Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-1-Phenylpropylamine 98.0+%, TCI America™
SDP

Catalog No. P15085G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P15085G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P15085G Supplier TCI America Supplier No. P15085G

missing translation for 'validMainframeMsg'

Specifications

CAS 3789-59-1
Melting Point -69°C
Color Yellow
Molecular Formula C9H13N
Quantity 5 g
MDL Number MFCD00082356
UN Number 2735
Synonym s-1-phenylpropan-1-amine, s---1-amino-1-phenylpropane, 1s-1-phenylpropan-1-amine, s---1-phenylpropylamine, s-1-phenylpropylamine, s---alpha-ethylbenzylamine, s---methylbenzylamine, 1-phenyl-propylamine, s---, a-ethylbenzylamine, pubchem6025
InChI Key AQFLVLHRZFLDDV-VIFPVBQESA-N
SMILES CCC(C1=CC=CC=C1)N
IUPAC Name (1S)-1-phenylpropan-1-amine
Molecular Weight (g/mol) 135.21
PubChem CID 6993773
Formula Weight 135.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (S)-(-)-1-Phenylpropylamine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.