Learn More
Medchemexpress LLC S-1-Propenyl-L-cysteine | 52438-09-2 | 99.9% | 161.22 | 10 MG
SDP
Catalog No. 5000436872
Encompass_Preferred
S-1-Propenyl-L-cysteine is a stereoisomer of S-allyl-l-cysteine, which is extracted from garlic. It demonstrates immunomodulatory effects and has been shown to reduce blood pressure in a hypertensive animal model. The compound also exhibits antioxidative efficacy through a nitric oxide (NO)-dependent BACH1 signaling pathway and is orally active.
  • Immunomodulatory effects: can modulate the immune system.
  • Antihypertensive properties: reduces blood pressure in hypertensive animal models.
  • Antioxidative efficacy: works through a NO-dependent BACH1 signaling pathway.
  • Oral activity: can be administered orally.
Catalog No. 50-004-36872 Supplier Medchemexpress LLC Supplier No. HY11182710MG
S-1-Propenyl-L-cysteine is a stereoisomer of S-allyl-l-cysteine, which is extracted from garlic. It demonstrates immunomodulatory effects and has been shown to reduce blood pressure in a hypertensive animal model. The compound also exhibits antioxidative efficacy through a nitric oxide (NO)-dependent BACH1 signaling pathway and is orally active.
  • Immunomodulatory effects: can modulate the immune system.
  • Antihypertensive properties: reduces blood pressure in hypertensive animal models.
  • Antioxidative efficacy: works through a NO-dependent BACH1 signaling pathway.
  • Oral activity: can be administered orally.