Learn More
Medchemexpress LLC S-1-Propenyl-L-cysteine | 52438-09-2 | 99.9% | 161.22 | 50 MG
SDP
Catalog No. 5000438918
Encompass_Preferred
S-1-Propenyl-L-cysteine is a stereoisomer of S-allyl-l-cysteine, extracted from garlic. It exhibits immunomodulatory effects and reduces blood pressure in a hypertensive animal model. It also shows antioxidative efficacy through a NO-dependent BACH1 signaling pathway. This compound is orally active.
  • Exhibits immunomodulatory effects
  • Reduces blood pressure in a hypertensive animal model
  • Shows antioxidative efficacy through a NO-dependent BACH1 signaling pathway
  • Is orally active
  • NO-dependently upregulates levels of HMOX1, causing downregulation and degradation of BACH1 in the nucleus
  • Inhibits lipopolysaccharides (LPS)-induced TLR4 signaling pathway by inducing MyD88 degradation
  • Reduces systolic blood pressure (SBP) and alters histidine metabolism in spontaneously hypertensive rats, exerting antihypertensive effects via the central histamine H3 receptor
Catalog No. 50-004-38918 Supplier Medchemexpress LLC Supplier No. HY11182750MG
S-1-Propenyl-L-cysteine is a stereoisomer of S-allyl-l-cysteine, extracted from garlic. It exhibits immunomodulatory effects and reduces blood pressure in a hypertensive animal model. It also shows antioxidative efficacy through a NO-dependent BACH1 signaling pathway. This compound is orally active.
  • Exhibits immunomodulatory effects
  • Reduces blood pressure in a hypertensive animal model
  • Shows antioxidative efficacy through a NO-dependent BACH1 signaling pathway
  • Is orally active
  • NO-dependently upregulates levels of HMOX1, causing downregulation and degradation of BACH1 in the nucleus
  • Inhibits lipopolysaccharides (LPS)-induced TLR4 signaling pathway by inducing MyD88 degradation
  • Reduces systolic blood pressure (SBP) and alters histidine metabolism in spontaneously hypertensive rats, exerting antihypertensive effects via the central histamine H3 receptor