Learn More
Medchemexpress LLC HY-111827 5mg , S-1-Propenyl-L-cysteine CAS:52438-09-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1118275MG
Encompass_Preferred
HY-111827 5mg S-1-Propenyl-L-cysteine CAS: 52438-09-2 S-1-Propenyl-L-cysteine is a stereoisomer of S-allyl-l-cysteine, extracted from garlic, with immunomodulatory effects and reduces blood pressure in a hypertensive animal model. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1659