Learn More
Strem, An Ascensus Company CAS# 649559-66-0. 500mg. (S)-(+)-1-[(R)-2-(Di-2-furylphosphino)ferrocenyl]ethyldi-3,5-xylylphosphine, min. 97%. MFCD06658128
SDP
Supplier:  Strem, An Ascensus Company 261170500MG
Encompass
CAS# 649559-66-0. 500mg. (S)-(+)-1-[(R)-2-(Di-2-furylphosphino)ferrocenyl]ethyldi-3,5-xylylphosphine, min. 97%. MFCD06658128. Molecular Weight: 618.46. Molecular Formula: C36H36FeO2P2. Color/form: orange pwdr. Strem# 26-1170. http://www.strem.com/catalog/v/26-1170/
Catalog No. 50-901-11032