Learn More
Medchemexpress LLC (S)-1-((S)-2-Amino-4-methylpentanoyl)pyrrolidine-2-carboxylic acid hydrochloride | 87178-63-0 | 98.25% | 264.75 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW01173050MG
Encompass_Preferred
Leucylproline hydrochloride is a dipeptide with a molecular weight of 264.75 and a purity of 98.25%. It is intended for research use only and has not been fully validated for medical applications.
  • White to off-white solid appearance
  • Consistent with structure by analytical methods
  • For research use only
  • Store powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
Catalog No. 50-004-52601