Learn More
Medchemexpress LLC (S)-1-t-butyl 2-methyl 4-oxopiperidine-1,2-dicarboxylate | 756486-14-3 | 97.0% | 257.29 g/mol | C12H19NO5 | 100 MG
SDP
Catalog No. 5000210870
Encompass_Preferred
AN-12-H5 intermediate-2 is a synthetic chemical intermediate used to prepare the viral inhibitor AN-12-H5 and as a building block for antibody-drug conjugates (ADCs). The compound is supplied as a solid with reported purity of 97.0%, molecular formula C12H19NO5, and molecular weight 257.29 g/mol.
  • Used as a synthetic intermediate for antiviral compound synthesis.
  • Suitable for antibody-drug conjugate (ADC) synthesis.
  • Reported purity 97.0% (typical analytical grade).
  • Solid form; recommended storage -20°C.
Catalog No. 50-002-10870 Supplier Medchemexpress LLC Supplier No. HYW047496100MG
AN-12-H5 intermediate-2 is a synthetic chemical intermediate used to prepare the viral inhibitor AN-12-H5 and as a building block for antibody-drug conjugates (ADCs). The compound is supplied as a solid with reported purity of 97.0%, molecular formula C12H19NO5, and molecular weight 257.29 g/mol.
  • Used as a synthetic intermediate for antiviral compound synthesis.
  • Suitable for antibody-drug conjugate (ADC) synthesis.
  • Reported purity 97.0% (typical analytical grade).
  • Solid form; recommended storage -20°C.