Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-1-(tert-Butoxycarbonyl)-2-azetidinemethanol 95.0+%, TCI America™
SDP

Catalog No. B21231G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21231G 1 g
B2123100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B21231G Supplier TCI America Supplier No. B21231G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 161511-85-9
Molecular Formula C9H17NO3
Molecular Weight (g/mol) 187.24
MDL Number MFCD03093620
InChI Key XIRUXUKRGUFEKC-UHFFFAOYNA-N
Synonym (S)-1-Boc-2-azetidinemethanol
PubChem CID 10511797
IUPAC Name tert-butyl 2-(hydroxymethyl)azetidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC1CO

Specifications

Color Yellow
Quantity 1 g
Formula Weight 187.24
Percent Purity ≥95.0% (GC)
Grade Research
Physical Form Liquid
Chemical Name or Material (S)-1-(tert-Butoxycarbonyl)-2-azetidinemethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.