Learn More
Medchemexpress LLC (S)-1-tert-butyl 2-methyl 4-oxopiperidine-1,2-dicarboxylate | 756486-14-3 | MFCD12547027 | 97.0% | 257.28 g/mol | C12H19NO5 | 250 MG
SDP
Catalog No. 5000209678
Encompass_Preferred
AN-12-H5 intermediate-2 (CAS 756486-14-3) is a synthetic small-molecule intermediate used in the synthesis of the viral inhibitor AN-12-H5 and as a building block for antibody-drug conjugate (ADC) development. It is supplied as a solid with reported purity ≥97.0% and is intended for research and medicinal chemistry applications.
  • High purity: ≥97.0%.
  • Solid form supplied in small research pack sizes.
  • Intended application: intermediate for small-molecule synthesis and ADC preparation.
  • Typical pack sizes: 50 mg-1 g (research quantities).
  • Storage guidance: pure form typically stored at -20°C; in solvent at -80°C.
Catalog No. 50-002-09678 Supplier Medchemexpress LLC Supplier No. 250MGHYW047496
AN-12-H5 intermediate-2 (CAS 756486-14-3) is a synthetic small-molecule intermediate used in the synthesis of the viral inhibitor AN-12-H5 and as a building block for antibody-drug conjugate (ADC) development. It is supplied as a solid with reported purity ≥97.0% and is intended for research and medicinal chemistry applications.
  • High purity: ≥97.0%.
  • Solid form supplied in small research pack sizes.
  • Intended application: intermediate for small-molecule synthesis and ADC preparation.
  • Typical pack sizes: 50 mg-1 g (research quantities).
  • Storage guidance: pure form typically stored at -20°C; in solvent at -80°C.