Learn More
Apexbio Technology LLC (S)-10-Hydroxycamptothecin, 25mg. Cas: 19685-09-7 MFCD: MFCD00189425. inhibitor of topoisomerase I
SDP
Supplier:  Apexbio Technology LLC B474525
Encompass_Preferred
IC50: 0.31 M10-Hydroxycamptothecin is a dose-dependent growth inhibitor of human microvascular endothelial cells (HMEC), and significantly inhibits the migration of HMEC with IC50 of 0.63 M, resulting in a dose-dependent inhibition of tube formation with IC50 of 0.96 M [1,2]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5357