Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-10-Methyl-1(9)-octal-2-one 98.5%, Thermo Scientific™

Catalog No. AC240802500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC240802500 250 mg
AC240800010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC240802500 Supplier Thermo Scientific Chemicals Supplier No. 240802500

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4087-39-2
Molecular Formula C11H16O
Molecular Weight (g/mol) 164.248
InChI Key OHERZLWVBJCXOF-NSHDSACASA-N
Synonym s-+-10-methyl-1 9-octal-2-one, 4as-4a-methyl-3,4,5,6,7,8-hexahydronaphthalen-2-one, 4a-methyl-4,4a,5,6,7,8-hexahydro-2 3h-naphthalenone #, 2 3h-naphthalenone, 4,4a,5,6,7,8-hexahydro-4a.beta.-methyl, 2 3h-naphthalenone, 4,4a,5,6,7,8-hexahydro-4a-methyl-, s, s-+-4,4a,5,6,7,8-hexahydro-4a-methyl-2 3h-naphthalenone, s-+-4a-methyl-2,3,4,4a,5,6,7,8-octahydro-2-naphthalenone, s-4,4a,5,6,7,8-hexahydro-4abeta-methyl-2 3h-naphthalenone, s-+-4,4a,5,6,7,8-hexahydro-4a-methyl-2 3h-naphthalenone, purum sum of enantiomers, gc
PubChem CID 7000123
IUPAC Name (4aS)-4a-methyl-3,4,5,6,7,8-hexahydronaphthalen-2-one
SMILES CC12CCCCC1=CC(=O)CC2

Specifications

CAS Min % 98.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.01
Infrared Spectrum Authentic
Refractive Index 1.5235 to 1.5255 (20°C, 589nm)
Quantity 250 mg
Specific Rotation +202° to +212° (20°C, 589nm) (c=1, ethanol)
Formula Weight 164.25
Percent Purity ≥98.0%
Physical Form Liquid
Chemical Name or Material (S)-(+)-10-Methyl-1(9)-octal-2-one, 98.5%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.