Learn More
Advanced ImmunoChemical, Inc. S-100 Protein, Ββ Homodimer, Human, Source - Human Brain Tissue
Supplier:  Advanced ImmunoChemical, Inc. 8S100BBH0.1MG
Encompass_Preferred
This item is manufactured by HyTest, Ltd and distributed by Advanced ImmunoChemical, Inc. Antigens / Neuroscience / S-100 protein, ββ homodimer, human. Human brain tissue.Blood samples from tissue donors were tested and found to be negative for HBsAg, HIV-1 and HIV-2 antibodies, HCV, and syphilis. S-100 – ββ homodimer (S100b) is suitable for use as a standard in immunoassay, as an immunogen for antiserum production and tracer for iodination. Measurements of S-100 protein in serumsamples of patients are useful in monitoring of traumatic brain injury, ischemic brain damage after circulatory arrests, in diagnosis and prognosis of clinical outcome in acute stroke. Purity > 95 %. After native gel electrophoresis by Ornstein-Davis S-100b protein is presented by one band ββ-form.S-100 protein concentration was determined by Lowry method using BSA as a standard. Lyophilized from 5 mM Tris-HCl, pH 7.5 with 5 mM 2-mercaptoethanol and 2 mM EDTA.
Catalog No. 50-153-9855