Learn More
Medchemexpress LLC S-14506 hydrochloride | 286369-38-8 | 99.9% | 443.94 g/mol | C24H27ClFN3O2 | 5 MG
SDP
Catalog No. 5000263861
Encompass_Preferred
S-14506 hydrochloride is the hydrochloride salt of S-14506, a potent 5-HT1A receptor agonist that also shows 5-HT2A/2C antagonist and dopamine antagonist activity. It is supplied as a high-purity research reagent for preclinical pharmacology, receptor-binding, and cell-based assays, available as solid material and DMSO solutions.
  • Potent 5-HT1A agonist with 5-HT2A/2C antagonist properties.
  • Also displays dopamine antagonist activity.
  • High purity (99.92%), suitable for analytical and biological assays.
  • Available as solid and 10 mM DMSO solution formats.
  • Supplied in small research pack sizes (for example, 5 mg).
Catalog No. 50-002-63861 Supplier Medchemexpress LLC Supplier No. HY1100245MG
S-14506 hydrochloride is the hydrochloride salt of S-14506, a potent 5-HT1A receptor agonist that also shows 5-HT2A/2C antagonist and dopamine antagonist activity. It is supplied as a high-purity research reagent for preclinical pharmacology, receptor-binding, and cell-based assays, available as solid material and DMSO solutions.
  • Potent 5-HT1A agonist with 5-HT2A/2C antagonist properties.
  • Also displays dopamine antagonist activity.
  • High purity (99.92%), suitable for analytical and biological assays.
  • Available as solid and 10 mM DMSO solution formats.
  • Supplied in small research pack sizes (for example, 5 mg).