Learn More
Medchemexpress LLC (S)-17-(4-(((2-Amino-4-oxo-3,4-dihydropteridin-6-yl)methyl)amino)benzamido)-14-oxo-4,7,10-trioxa-13-azaoctadecane-1,18-dioic acid | 97.6% | C28H36N8O10 | 5 MG
SDP
Catalog No. 5000358282
Encompass_Preferred
Folate-PEG3-C2-acid is an acid fragment and PEG-type PROTAC linker used in the synthesis of Proteolysis-targeting Chimeras. It has a molecular weight of 644.63 and a purity of 97.56%.
  • Stable under recommended storage conditions
  • Consistent with structure based on 1H NMR Spectrum and LCMS
  • Solid appearance
  • For research use only
Catalog No. 50-003-58282 Supplier Medchemexpress LLC Supplier No. HY473735MG
Folate-PEG3-C2-acid is an acid fragment and PEG-type PROTAC linker used in the synthesis of Proteolysis-targeting Chimeras. It has a molecular weight of 644.63 and a purity of 97.56%.
  • Stable under recommended storage conditions
  • Consistent with structure based on 1H NMR Spectrum and LCMS
  • Solid appearance
  • For research use only