Learn More
Medchemexpress LLC S 17092 | 176797-26-5 | MFCD00948255 | 99.8% | 384.53 g·mol⁻¹ | C22H28N2O2S | 10 MG
SDP
Catalog No. 5000285913
Encompass_Preferred
S 17092 is an orally active small-molecule prolyl endopeptidase (PEP) inhibitor provided for research use. It has reported high potency against PEP and is supplied in both solid and solution formats for use in biochemical and preclinical studies.
  • High biochemical potency (IC50 1.2 nM).
  • High purity (≈99.8%) for consistent assay performance.
  • Available as solid powder and as a 10 mM solution in DMSO to suit assay needs.
  • Comes with datasheet, certificate of analysis, and safety data sheet for quality and safety verification.
  • Recommended storage conditions provided to maintain compound stability.
  • Suitable for in vitro biochemical and cellular studies targeting prolyl endopeptidase.
Catalog No. 50-002-85913 Supplier Medchemexpress LLC Supplier No. HY1511410MG
S 17092 is an orally active small-molecule prolyl endopeptidase (PEP) inhibitor provided for research use. It has reported high potency against PEP and is supplied in both solid and solution formats for use in biochemical and preclinical studies.
  • High biochemical potency (IC50 1.2 nM).
  • High purity (≈99.8%) for consistent assay performance.
  • Available as solid powder and as a 10 mM solution in DMSO to suit assay needs.
  • Comes with datasheet, certificate of analysis, and safety data sheet for quality and safety verification.
  • Recommended storage conditions provided to maintain compound stability.
  • Suitable for in vitro biochemical and cellular studies targeting prolyl endopeptidase.