Learn More
Medchemexpress LLC HY-10936 10mg Medchemexpress, S 18986 CAS:175340-20-2 Purity:>98%
SDP
Catalog No. 501968696
Encompass_Preferred
Medchemexpress, HY-10936 10mg S 18986 CAS:175340-20-2 S 18986 is a selective, orally active, brain penetrant positive allosteric modulator of AMPA-type receptors. S 18986 shows cognitive enhancing properties in rodents. S 18986 activates the release of noradrenaline and acetylcholine in rat hippocampus and enhances rat memory in object-recognition tests. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8696 Supplier Medchemexpress LLC Supplier No. HY1093610MG
Medchemexpress, HY-10936 10mg S 18986 CAS:175340-20-2 S 18986 is a selective, orally active, brain penetrant positive allosteric modulator of AMPA-type receptors. S 18986 shows cognitive enhancing properties in rodents. S 18986 activates the release of noradrenaline and acetylcholine in rat hippocampus and enhances rat memory in object-recognition tests. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.