Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S)-2-(1-(4-amino-3-(3-fluoro-4-methoxyphenyl)-1H-pyrazolo[3,4-d]pyrimidin-1-yl)propyl)-3-cyclopropyl-5- | 2429889-61-0 | 503.50 g/mol | C26H23F2N7O2 | 5 MG
SDP

Catalog No. 5000262667
Encompass_Preferred

IHMT-PI3Kδ-372 is a potent, selective PI3Kδ inhibitor research compound used in preclinical pharmacology and COPD-related studies. It exhibits nanomolar potency and high selectivity versus other class I PI3Ks, suitable for in vitro and in vivo research applications.

  • Potency: IC50 14 nM against PI3Kδ.
  • High selectivity over other class I PI3Ks.
  • Suitable for preclinical COPD and pharmacology research.
  • Molecular formula C26H23F2N7O2.
  • Molecular weight 503.50 g/mol.
  • Available in small milligram pack sizes for laboratory use.

Catalog No. 50-002-62667 Supplier Medchemexpress LLC Supplier No. HY1319105MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

IHMT-PI3Kδ-372 is a potent, selective PI3Kδ inhibitor research compound used in preclinical pharmacology and COPD-related studies. It exhibits nanomolar potency and high selectivity versus other class I PI3Ks, suitable for in vitro and in vivo research applications.

  • Potency: IC50 14 nM against PI3Kδ.
  • High selectivity over other class I PI3Ks.
  • Suitable for preclinical COPD and pharmacology research.
  • Molecular formula C26H23F2N7O2.
  • Molecular weight 503.50 g/mol.
  • Available in small milligram pack sizes for laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.