Learn More
Medchemexpress LLC (S)-2-(1,4-dibenzylpiperazin-2-yl)acetonitrile | 2225787-92-6 | 99.9% | 305.43 g·mol⁻¹ | C20H23N3 | 250 MG
SDP
Catalog No. 5000209408
Encompass_Preferred
(S)-2-(1,4-dibenzylpiperazin-2-yl)acetonitrile is a chiral small-molecule intermediate used in medicinal chemistry and organic synthesis to prepare bioactive compounds and analogs. It is supplied as a high-purity material intended for research and development applications.
  • Chiral (S)-enantiomer for stereoselective synthesis.
  • High purity: 99.9% (LCMS) for reliable reaction outcomes.
  • Molecular formula C20H23N3, molecular weight 305.43 g·mol⁻¹.
  • Commonly used as a synthetic building block and drug intermediate.
  • Supplied in a 250 mg quantity suitable for lab-scale synthesis.
  • Accompanied by Certificate of Analysis and safety data documentation.
Catalog No. 50-002-09408 Supplier Medchemexpress LLC Supplier No. 250MGHYW049455
(S)-2-(1,4-dibenzylpiperazin-2-yl)acetonitrile is a chiral small-molecule intermediate used in medicinal chemistry and organic synthesis to prepare bioactive compounds and analogs. It is supplied as a high-purity material intended for research and development applications.
  • Chiral (S)-enantiomer for stereoselective synthesis.
  • High purity: 99.9% (LCMS) for reliable reaction outcomes.
  • Molecular formula C20H23N3, molecular weight 305.43 g·mol⁻¹.
  • Commonly used as a synthetic building block and drug intermediate.
  • Supplied in a 250 mg quantity suitable for lab-scale synthesis.
  • Accompanied by Certificate of Analysis and safety data documentation.