Learn More
Medchemexpress LLC (S)-2-(2-Amino-2-methylpropanamido)-N-((3R,4S,5S)-3-methoxy-1-((S)-2-((1R,2R)-1-methoxy-2-methyl-3-oxo-3-(((S)-2-phenyl-1-(thiazol-2-yl)ethyl)amino)propyl)pyrrolidin-1-yl)-5-methyl-1-oxoheptan-4-yl)-N-methyl-3-(methyl-d3)butanamide-2,3,4,4,4-d5 | 1436391-86-4 | 99.3% | 751.06 | 25 MG
SDP
Catalog No. 5000440048
Encompass_Preferred
PF-06380101-d8 is a deuterium-labeled PF-06380101, which is an Auristatin microtubule inhibitor and a cytotoxic Dolastatin 10 analogue. This compound can be used as a tracer and an internal standard for quantitative analysis in various applications.
  • Deuterium labeled compound.
  • Auristatin microtubule inhibitor.
  • Cytotoxic dolastatin 10 analogue.
  • Stable heavy isotopes are incorporated for use as tracers for quantitation during drug development.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-40048 Supplier Medchemexpress LLC Supplier No. 25MGHY12522S
PF-06380101-d8 is a deuterium-labeled PF-06380101, which is an Auristatin microtubule inhibitor and a cytotoxic Dolastatin 10 analogue. This compound can be used as a tracer and an internal standard for quantitative analysis in various applications.
  • Deuterium labeled compound.
  • Auristatin microtubule inhibitor.
  • Cytotoxic dolastatin 10 analogue.
  • Stable heavy isotopes are incorporated for use as tracers for quantitation during drug development.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.