Learn More
Strem, An Ascensus Company CAS# 100165-88-6. (S)-(-)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl, 98% (S)-(-)-TolBINAP. MFCD01311709. 1G
SDP
Catalog No. 5090109801
Encompass
CAS# 100165-88-6. 1G. (S)-(-)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl, 98% (S)-(-)-TolBINAP. MFCD01311709. Molecular Formula: C48H40P2 Molecular Weight: 678.79. Color/Form: white pwdr. Strem#: 15-0153 www.strem.com/catalog/v3/15-0153/
Catalog No. 50-901-09801 Supplier Strem, An Ascensus Company Supplier No. 1501531G
CAS# 100165-88-6. 1G. (S)-(-)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl, 98% (S)-(-)-TolBINAP. MFCD01311709. Molecular Formula: C48H40P2 Molecular Weight: 678.79. Color/Form: white pwdr. Strem#: 15-0153 www.strem.com/catalog/v3/15-0153/