Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl, 98+%, ACROS Organics™

Click to view available options
Quantity:
250mg
1g
Packaging:
Glass bottle
100165-88-6
C48H40P2
678.796
MFCD01311709
IOPQYDKQISFMJI-UHFFFAOYSA-N
r-tol-binap,s-tol-binap,s---2,2'-bis di-p-tolylphosphino-1,1'-binaphthyl,2,2'-bis di-p-tolylphosphino-1,1'-binaphthyl,s-t-binap,rac-tol-binap,r-+-2,2'-bis di-p-tolylphosphino-1,1'-binaphthyl,tol-binap,s-2,2'-bis di-p-tolylphosphino-1,1'-binaphthyl,s-p-tol-binap
4189906
[1-[2-bis(4-methylphenyl)phosphanylnaphthalen-1-yl]naphthalen-2-yl]-bis(4-methylphenyl)phosphane
CC1=CC=C(C=C1)P(C2=CC=C(C=C2)C)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=C(C=C7)C)C8=CC=C(C=C8)C
100165-88-6
C48H40P2
678.796
MFCD01311709
IOPQYDKQISFMJI-UHFFFAOYSA-N
r-tol-binap,s-tol-binap,s---2,2'-bis di-p-tolylphosphino-1,1'-binaphthyl,2,2'-bis di-p-tolylphosphino-1,1'-binaphthyl,s-t-binap,rac-tol-binap,r-+-2,2'-bis di-p-tolylphosphino-1,1'-binaphthyl,tol-binap,s-2,2'-bis di-p-tolylphosphino-1,1'-binaphthyl,s-p-tol-binap
4189906
[1-[2-bis(4-methylphenyl)phosphanylnaphthalen-1-yl]naphthalen-2-yl]-bis(4-methylphenyl)phosphane
CC1=CC=C(C=C1)P(C2=CC=C(C=C2)C)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=C(C=C7)C)C8=CC=C(C=C8)C
Specific Rotation -175° to -185° (20°C, 589nm) (C=0,3, toluene)
Packaging Glass bottle
Quantity 1g
Color White
Infrared Spectrum Authentic
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Formula Weight 678.79
Physical Form Powder
Percent Purity ≥98.0%
Chemical Name or Material (S)-(-)-2, 2'-Bis(di-p-tolylphosphino)-1, 1'-binaphthyl
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.