Learn More
Strem, An Ascensus Company CAS# 133545-17-2. 100mg. (S)-(-)-2,2'-Bis(diphenylphosphino)-6,6'-dimethoxy-1,1'-biphenyl, min. 97% (S)-MeO-BIPHEP. MFCD03095430
SDP
Supplier:  Strem, An Ascensus Company 150179100MG
Encompass
CAS# 133545-17-2. 100mg. (S)-(-)-2,2'-Bis(diphenylphosphino)-6,6'-dimethoxy-1,1'-biphenyl, min. 97% (S)-MeO-BIPHEP. MFCD03095430. Molecular Weight: 582.53. Molecular Formula: C38H32O2P2. Color/form: white xtl. Strem# 15-0179. http://www.strem.com/catalog/v/15-0179/
Catalog No. 50-901-09828