Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-2,2'-Dimethyl-1,1'-binaphthyl 98.0+%, TCI America™
SDP

Catalog No. D28385G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D28385G 5 g
D28381G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D28385G Supplier TCI America Supplier No. D28385G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 32587-64-7
Molecular Formula C14H14
Molecular Weight (g/mol) 182.266
MDL Number MFCD00048075
InChI Key ABMKWMASVFVTMD-UHFFFAOYSA-N
PubChem CID 11797
IUPAC Name 1-methyl-2-(2-methylphenyl)benzene
SMILES CC1=CC=CC=C1C2=CC=CC=C2C

Specifications

Quantity 5 g
Formula Weight 282.39
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (S)-2,2′-Dimethyl-1,1′-binaphthyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.