Learn More
Medchemexpress LLC (S)-2-(2-((tert-butoxycarbonyl)amino)propanamido)acetic acid | 28782-78-7 | MFCD00235759 | 97.0% | 246.26 g/mol | C10H18N2O5 | 5 G
SDP
Catalog No. 5000299964
Encompass_Preferred
(S)-2-(2-((tert-Butoxycarbonyl)amino)propanamido)acetic acid is a Boc-protected glycine derivative used as a peptide fragment and building block in medicinal chemistry and peptide synthesis. It is supplied as a white to off-white solid with formula C10H18N2O5 and molecular weight 246.26 g/mol; CAS 28782-78-7. Typical applications include incorporation into peptides, synthetic modification, and structure-activity studies.
  • Boc-protected amino acid derivative for incorporation into peptides.
  • High purity suitable for research use (97.0%).
  • Solid, white to off-white form for straightforward handling.
  • Stable under recommended storage conditions to preserve integrity.
  • Useful as a building block in medicinal chemistry and peptide synthesis.
Catalog No. 50-002-99964 Supplier Medchemexpress LLC Supplier No. HYW0196845G
(S)-2-(2-((tert-Butoxycarbonyl)amino)propanamido)acetic acid is a Boc-protected glycine derivative used as a peptide fragment and building block in medicinal chemistry and peptide synthesis. It is supplied as a white to off-white solid with formula C10H18N2O5 and molecular weight 246.26 g/mol; CAS 28782-78-7. Typical applications include incorporation into peptides, synthetic modification, and structure-activity studies.
  • Boc-protected amino acid derivative for incorporation into peptides.
  • High purity suitable for research use (97.0%).
  • Solid, white to off-white form for straightforward handling.
  • Stable under recommended storage conditions to preserve integrity.
  • Useful as a building block in medicinal chemistry and peptide synthesis.