Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-2,3,7,7a-Tetrahydro-7a-methyl-1H-indene-1,5(6H)-dione, 90%

Catalog No. p-3830592
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC296470010 1 g Glass bottle
AC296470050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC296470010 Supplier Thermo Scientific Chemicals Supplier No. 296470010
Only null left
Add to Cart

CAS: 17553-86-5 | C10H12O2 | 164.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17553-86-5
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.2
MDL Number MFCD00012272
InChI Key FNYAZSZTENLTRT-JTQLQIEISA-N
Synonym 7as-7a-methyl-2,3,6,7-tetrahydroindene-1,5-dione, s-+-2,3,7,7a-tetrahydro-7a-methyl-1h-indene-1,5 6h-dione, 1h-indene-1,5 6h-dione, 2,3,7,7a-tetrahydro-7a-methyl-, 7as, hajos dione, s-hajos dione, s-hajos ketone, s-hajos-wiechert ketone, +-hajos-parrish ketone, +-hajos-parrish diketone, 7as-7a-methyl-2,3,7,7a-tetrahydro-1h-indene-1,5 6h-dione
PubChem CID 736943
IUPAC Name (7aS)-7a-methyl-2,3,6,7-tetrahydroindene-1,5-dione
SMILES CC12CCC(=O)C=C1CCC2=O

Specifications

CAS Min % 88.0
CAS Max % 100.0
Melting Point 55.0°C to 64.0°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Quantity 1 g
Specific Rotation Condition +331° (20°C c=1,toluene)
Specific Rotation + 331.00
Formula Weight 164.2
Percent Purity 90%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material (S)-(+)-2,3,7,7a-Tetrahydro-7a-methyl-1H-indene-1,5(6H)-dione
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.