Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S)-2,3-bis((tert-butoxycarbonyl)amino)propanoic acid | 88971-40-8 | MFCD00144826 | ≥95.0% | 304.34 g/mol | C13H24N2O6 | 1 G
SDP

Catalog No. 5000209883
Encompass_Preferred

Boc-Dap(Boc)-OH is a Boc-protected diaminopropionic acid used as a protected amino acid building block for peptide synthesis. It is supplied as a solid and commonly used in solid-phase and solution-phase peptide assembly.

  • Suitable for solid-phase and solution-phase peptide synthesis
  • Systematic name: (S)-2,3-bis((tert-butoxycarbonyl)amino)propanoic acid
  • Purity ≥95.0% (NMR assay)
  • Molecular weight 304.34 g/mol, formula C13H24N2O6
  • Storage: room temperature (solid); in solvent store at -20°C to -80°C

Catalog No. 50-002-09883 Supplier Medchemexpress LLC Supplier No. HYW0477991G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boc-Dap(Boc)-OH is a Boc-protected diaminopropionic acid used as a protected amino acid building block for peptide synthesis. It is supplied as a solid and commonly used in solid-phase and solution-phase peptide assembly.

  • Suitable for solid-phase and solution-phase peptide synthesis
  • Systematic name: (S)-2,3-bis((tert-butoxycarbonyl)amino)propanoic acid
  • Purity ≥95.0% (NMR assay)
  • Molecular weight 304.34 g/mol, formula C13H24N2O6
  • Storage: room temperature (solid); in solvent store at -20°C to -80°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.