Learn More
Medchemexpress LLC S-(2,4-Dinitrophenyl)glutathione | 26289-39-4 | 99.89% | 473.41 | 1 MG
SDP
Catalog No. 5000348570
Encompass_Preferred
S-(2,4-Dinitrophenyl)glutathione is a substrate for glutathione-S-transferase. It can also function as an irreversible glutathione reductase inhibitor, with a Ki value of 30 μM.
  • Substrate for glutathione-S-transferase
  • Irreversible glutathione reductase inhibitor
  • Ki value of 30 μM
Catalog No. 50-003-48570 Supplier Medchemexpress LLC Supplier No. HY1373301MG
S-(2,4-Dinitrophenyl)glutathione is a substrate for glutathione-S-transferase. It can also function as an irreversible glutathione reductase inhibitor, with a Ki value of 30 μM.
  • Substrate for glutathione-S-transferase
  • Irreversible glutathione reductase inhibitor
  • Ki value of 30 μM