Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC S-(2,4-dinitrophenyl)glutathione | 26289-39-4 | MFCD01863384 | 99.9% | 473.41 g/mol | C16H19N5O10S | 5 MG
SDP

Catalog No. 5000222419
Encompass_Preferred

S-(2,4-dinitrophenyl)glutathione is a glutathione derivative in which the cysteine thiol is modified with a 2,4-dinitrophenyl group. It is used as a substrate and inhibitor in biochemical studies of glutathione-related enzymes and for enzymatic assays, supplied as a solid with analytical documentation for research use.

  • High purity (99.89%).
  • Molecular weight 473.41 g/mol.
  • Molecular formula C16H19N5O10S.
  • Available sizes: 1 mg, 5 mg, 10 mg (larger sizes by request).
  • Storage: sealed powder at -80°C (long term) or -20°C (short term).
  • Datasheet, SDS, and COA available for quality and handling information.

Catalog No. 50-002-22419 Supplier Medchemexpress LLC Supplier No. HY1373305MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

S-(2,4-dinitrophenyl)glutathione is a glutathione derivative in which the cysteine thiol is modified with a 2,4-dinitrophenyl group. It is used as a substrate and inhibitor in biochemical studies of glutathione-related enzymes and for enzymatic assays, supplied as a solid with analytical documentation for research use.

  • High purity (99.89%).
  • Molecular weight 473.41 g/mol.
  • Molecular formula C16H19N5O10S.
  • Available sizes: 1 mg, 5 mg, 10 mg (larger sizes by request).
  • Storage: sealed powder at -80°C (long term) or -20°C (short term).
  • Datasheet, SDS, and COA available for quality and handling information.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.