Learn More
Medchemexpress LLC S-[2-[6-[[4-[3-(dimethylamino)propoxy]phenyl]sulfonylamino]-3-pyridinyl]-2-oxoethyl] ethanethioate | 940943-37-3 | MFCD19690928 | >96.0% | 451.6 g/mol | C20H25N3O5S2 | 50 MG
SDP
Catalog No. 5000240894
Encompass_Preferred
KD 5170 is a mercaptoketone-based pan inhibitor of histone deacetylases (HDACs) that exhibits broad-spectrum antitumor activity in vitro and in vivo. It is provided as a solid research reagent for biochemical and cellular studies of HDAC inhibition, apoptosis, and cell-cycle regulation.
  • Pan-HDAC inhibitor active against multiple HDAC isoforms.
  • Demonstrates potent biochemical and cellular activity.
  • Validated for in vitro and in vivo antitumor studies.
  • Supplied as a solid with high purity (>96%).
  • Chemical formula C20H25N3O5S2; molecular weight ~451.6 g/mol.
  • Intended for research use in oncology and epigenetics.
Catalog No. 50-002-40894 Supplier Medchemexpress LLC Supplier No. HY10754950MG
KD 5170 is a mercaptoketone-based pan inhibitor of histone deacetylases (HDACs) that exhibits broad-spectrum antitumor activity in vitro and in vivo. It is provided as a solid research reagent for biochemical and cellular studies of HDAC inhibition, apoptosis, and cell-cycle regulation.
  • Pan-HDAC inhibitor active against multiple HDAC isoforms.
  • Demonstrates potent biochemical and cellular activity.
  • Validated for in vitro and in vivo antitumor studies.
  • Supplied as a solid with high purity (>96%).
  • Chemical formula C20H25N3O5S2; molecular weight ~451.6 g/mol.
  • Intended for research use in oncology and epigenetics.