Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC S-[2-[6-[[4-[3-(dimethylamino)propoxy]phenyl]sulfonylamino]-3-pyridinyl]-2-oxoethyl] ethanethioate | 940943-37-3 | MFCD19690928 | 98.1% | 451.56 | C20H25N3O5S2 | 10 MG
SDP

Catalog No. 5000241454
Encompass_Preferred

KD 5170 is a small-molecule pan inhibitor of histone deacetylases (HDACs) that demonstrates broad-spectrum antitumor activity in vitro and in vivo. It is offered for laboratory research use and includes recommended solubility and storage guidance for experimental applications.

  • Pan inhibitor of histone deacetylases (HDACs).
  • Demonstrates broad-spectrum antitumor activity in vitro and in vivo.
  • High purity (≈98.1%).
  • Molecular formula C20H25N3O5S2; molecular weight 451.56.
  • Soluble in DMSO (100 mg/mL); in vivo formulations documented at 2.5 mg/mL in common vehicles.
  • Store sealed at 4°C; in solvent store at -80°C for long-term stability.

Catalog No. 50-002-41454 Supplier Medchemexpress LLC Supplier No. HY10754910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

KD 5170 is a small-molecule pan inhibitor of histone deacetylases (HDACs) that demonstrates broad-spectrum antitumor activity in vitro and in vivo. It is offered for laboratory research use and includes recommended solubility and storage guidance for experimental applications.

  • Pan inhibitor of histone deacetylases (HDACs).
  • Demonstrates broad-spectrum antitumor activity in vitro and in vivo.
  • High purity (≈98.1%).
  • Molecular formula C20H25N3O5S2; molecular weight 451.56.
  • Soluble in DMSO (100 mg/mL); in vivo formulations documented at 2.5 mg/mL in common vehicles.
  • Store sealed at 4°C; in solvent store at -80°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.