Learn More
Medchemexpress LLC S-[2-[6-[[4-[3-(dimethylamino)propoxy]phenyl]sulfonylamino]-3-pyridinyl]-2-oxoethyl] ethanethioate | 940943-37-3 | MFCD19690928 | 98.1% | 451.56 g·mol⁻¹ | C20H25N3O5S2 | 25 MG
SDP
Catalog No. 5000240833
Encompass_Preferred
KD 5170 is a mercaptoketone-based pan inhibitor of histone deacetylases (HDACs) that demonstrates broad-spectrum antitumor activity in vitro and in vivo. It is supplied as a white to yellow solid for research use only.
  • Potent pan-histone deacetylase (HDAC) inhibition.
  • Demonstrated broad-spectrum antitumor activity in vitro and in vivo.
  • High reported purity (≈98.1%).
  • Available in small-mass pack sizes suitable for preclinical studies.
  • White to yellow solid form, convenient for storage and handling.
  • Intended for research use only and not for human therapeutic use.
Catalog No. 50-002-40833 Supplier Medchemexpress LLC Supplier No. HY10754925MG
KD 5170 is a mercaptoketone-based pan inhibitor of histone deacetylases (HDACs) that demonstrates broad-spectrum antitumor activity in vitro and in vivo. It is supplied as a white to yellow solid for research use only.
  • Potent pan-histone deacetylase (HDAC) inhibition.
  • Demonstrated broad-spectrum antitumor activity in vitro and in vivo.
  • High reported purity (≈98.1%).
  • Available in small-mass pack sizes suitable for preclinical studies.
  • White to yellow solid form, convenient for storage and handling.
  • Intended for research use only and not for human therapeutic use.