Learn More
Medchemexpress LLC (S)-2,6-Bis((tert-butoxycarbonyl)amino)hexanoic acid | 2483-46-7 | 346.42 | 100 G
SDP
Catalog No. 5000448735
Encompass_Preferred
(S)-2,6-Bis((tert-butoxycarbonyl)amino)hexanoic acid is a polypeptide derivative utilized in various chemical syntheses. It serves as a key component in the creation of multifunctional amphiphilic peptide dendrimers, which are crucial for non-viral gene delivery systems in cancer research. Additionally, this compound is employed in the preparation of organic substances that enhance the luminescence intensity of alkaline phosphatase substrates, highlighting its versatility in biochemical applications.
  • Utilized in synthesizing amphiphilic peptide dendrimers
  • Supports non-viral gene delivery in cancer research
  • Enhances luminescence intensity of alkaline phosphatase substrates
Catalog No. 50-004-48735 Supplier Medchemexpress LLC Supplier No. HYW007599100G
(S)-2,6-Bis((tert-butoxycarbonyl)amino)hexanoic acid is a polypeptide derivative utilized in various chemical syntheses. It serves as a key component in the creation of multifunctional amphiphilic peptide dendrimers, which are crucial for non-viral gene delivery systems in cancer research. Additionally, this compound is employed in the preparation of organic substances that enhance the luminescence intensity of alkaline phosphatase substrates, highlighting its versatility in biochemical applications.
  • Utilized in synthesizing amphiphilic peptide dendrimers
  • Supports non-viral gene delivery in cancer research
  • Enhances luminescence intensity of alkaline phosphatase substrates