Learn More
Medchemexpress LLC (S)-2,6-Bis((tert-butoxycarbonyl)amino)hexanoic acid | 2483-46-7 | 346.42 | 25 G
SDP
Catalog No. 5000449266
Encompass_Preferred
(S)-2,6-Bis((tert-butoxycarbonyl)amino)hexanoic acid is a polypeptide derivative. It is used to synthesize multifunctional amphiphilic peptide dendrimers for non-viral gene delivery in cancer research and in the preparation of organic substances that enhance the luminescence intensity of alkaline phosphatase substrates.
  • Synthesizes nonviral gene vectors for loading and delivering DNA.
  • Synthesizes a luminous enhancement agent to improve the luminous intensity of alkaline phosphatase substrates and enhance the sensitivity of chemiluminescence detection.
Catalog No. 50-004-49266 Supplier Medchemexpress LLC Supplier No. 25GHYW007599
(S)-2,6-Bis((tert-butoxycarbonyl)amino)hexanoic acid is a polypeptide derivative. It is used to synthesize multifunctional amphiphilic peptide dendrimers for non-viral gene delivery in cancer research and in the preparation of organic substances that enhance the luminescence intensity of alkaline phosphatase substrates.
  • Synthesizes nonviral gene vectors for loading and delivering DNA.
  • Synthesizes a luminous enhancement agent to improve the luminous intensity of alkaline phosphatase substrates and enhance the sensitivity of chemiluminescence detection.