Learn More
Medchemexpress LLC (S)-2,6-Bis((tert-butoxycarbonyl)amino)hexanoic acid | 2483-46-7 | MFCD00038515 | 346.42 | 10 G
SDP
Catalog No. 5000446480
Encompass_Preferred
(S)-2,6-Bis((tert-butoxycarbonyl)amino)hexanoic acid is a polypeptide derivative. It is used in the synthesis of multifunctional amphiphilic peptide dendrimers for non-viral gene delivery in cancer research. Additionally, this compound can be utilized in the preparation of organic substances that enhance the luminescence intensity of alkaline phosphatase substrates, improving the sensitivity of chemiluminescence detection.
  • Functions as a polypeptide derivative
  • Supports non-viral gene delivery in cancer research
  • Enhances luminescence intensity of alkaline phosphatase substrates
Catalog No. 50-004-46480 Supplier Medchemexpress LLC Supplier No. HYW00759910G
(S)-2,6-Bis((tert-butoxycarbonyl)amino)hexanoic acid is a polypeptide derivative. It is used in the synthesis of multifunctional amphiphilic peptide dendrimers for non-viral gene delivery in cancer research. Additionally, this compound can be utilized in the preparation of organic substances that enhance the luminescence intensity of alkaline phosphatase substrates, improving the sensitivity of chemiluminescence detection.
  • Functions as a polypeptide derivative
  • Supports non-viral gene delivery in cancer research
  • Enhances luminescence intensity of alkaline phosphatase substrates