Learn More
Medchemexpress LLC (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)-4-oxo-4-(tritylamino)butanoic acid | 941296-80-6 | MFCD22200773 | 97.0% | 610.70 g·mol⁻¹ | C39H34N2O5 | 250 MG
SDP
Catalog No. 5000283121
Encompass_Preferred
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)-4-oxo-4-(tritylamino)butanoic acid is an Fmoc- and Trt-protected N-methyl asparagine derivative intended as a building block for peptide chemistry. It is supplied as a solid and is used to introduce Nα-methylated asparagine residues in solid-phase peptide synthesis and peptide modification studies.
  • Protected N-methyl asparagine for peptide synthesis.
  • Includes Fmoc and trityl protecting groups compatible with standard deprotection protocols.
  • Enables introduction of Nα-methyl residues to modify peptide conformation.
  • Supplied as a solid with defined storage recommendations to preserve stability.
  • Suitable for research-scale synthesis and method development.
Catalog No. 50-002-83121 Supplier Medchemexpress LLC Supplier No. HYW130397250MG
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)-4-oxo-4-(tritylamino)butanoic acid is an Fmoc- and Trt-protected N-methyl asparagine derivative intended as a building block for peptide chemistry. It is supplied as a solid and is used to introduce Nα-methylated asparagine residues in solid-phase peptide synthesis and peptide modification studies.
  • Protected N-methyl asparagine for peptide synthesis.
  • Includes Fmoc and trityl protecting groups compatible with standard deprotection protocols.
  • Enables introduction of Nα-methyl residues to modify peptide conformation.
  • Supplied as a solid with defined storage recommendations to preserve stability.
  • Suitable for research-scale synthesis and method development.