Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (S)-(-)-2'-Amino-1,1'-Binaphthalen-2-Ol

Catalog No. 713694100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-369-4100MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 71-369-4100MG Supplier Sigma Aldrich Supplier No. 713694100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(S)-(-)-2'sa-Amino-1,1'sa-binaphthalen-2-ol or (S)-NOBIN, is a non-symmetrically substituted 1,1'sa-binaphthalene ligand. It can be used as a catalyst in the asymmetric synthesis of unnatural +a-alkyl amino acids by phase-transfer catalysis (PTC). (S)-NOBIN can also be employed as starting material to prepare N-monoalkyl and N,N-dialkyl (S)-NOBIN analogs by reacting with aldehydes via reductive amination reaction. N-monoaryl derivatives are obtained from NOBIN via Hartwig-Buchwald arylation reaction.

Specifications

CAS 69861-71-8
Melting Point 220°C to 225°C
Molecular Formula C42H42P2Pd
Linear Formula C42H42P2Pd
MDL Number MFCD12911908
Quantity 500 mg
Synonym Bis(tris(2-tolyl)phosphine)palladium; Pd[(o-tol)3P]2
Molecular Weight (g/mol) 715.15 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.