Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (S)-(+)-2-Amino-1-Butanol

Catalog No. 1325275G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-252-75G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 13-252-75G Supplier Sigma Aldrich Supplier No. 1325275G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(S)-(+)-2-Amino-1-butanol is a chiral amino alcohol that may be used in the synthesis of (S)-2-(6-benzylamino-9-isopropyl-9H-purin-2-ylamino) butan-1-ol, an (S)-enantiomer of roscovitine. It can also be used to synthesize homochiral 2-methylpyrroles. (S)-(+)-2-Amino-1-butanol is an intermediate for the synthesis of (S,S)-ethambutol, an antibacterial agent for treating tuberculosis.

Specifications

CAS 14434-22-1
Molecular Formula C6FeN6Na4 -+ 10H2O
Refractive Index n20/D 1.4521
Linear Formula Na4Fe(CN)6 -+ 10H2O
MDL Number MFCD00149274
Quantity 500 g
Synonym Tetrasodium hexacyanoferrate decahydrate; Sodium hexacyanoferrate(II)
Molecular Weight (g/mol) 484.06 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.