Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-2-Amino-1-propanol, 98%

Catalog No. AC104450100
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC104450100 10 g Glass bottle
AC104450010 1 g Glass bottle
AC104450500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC104450100 Supplier Thermo Scientific Chemicals Supplier No. 104450100
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 2749-11-3 | C3H9NO | 75.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2749-11-3
Molecular Formula C3H9NO
Molecular Weight (g/mol) 75.11
MDL Number MFCD00064412
InChI Key BKMMTJMQCTUHRP-VKHMYHEASA-O
Synonym l-alaninol, s-+-2-amino-1-propanol, s-2-aminopropan-1-ol, 2s-2-aminopropan-1-ol, h-alaninol, alaninol, s-2-amino-1-propanol, +-2-aminopropan-1-ol, 2-aminopropanol, +, unii-v403gh89l1
PubChem CID 80307
ChEBI CHEBI:78502
IUPAC Name (2S)-2-aminopropan-1-ol
SMILES CC(CO)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9650g/mL
Boiling Point 173°C to 176°C
Flash Point 62°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.4480 to 1.451
Linear Formula CH3CH(NH2)CH2OH
Quantity 10 g
Beilstein 04, I, 432
Specific Gravity 0.965
Merck Index 10, 472
Specific Rotation + 22
Solubility Information Solubility in water: freely soluble. Other solubilities: freely soluble in alcohol and ether
Formula Weight 75.11
Percent Purity 98%
Physical Form Oily Liquid
Chemical Name or Material (S)-(+)-2-Amino-1-propanol
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 220-388-7

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.