Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S)-2-amino-2-(2-chlorophenyl)acetic acid | 141315-50-6 | MFCD03426346 | ≥97.0% | 185.61 g·mol⁻¹ | C8H8ClNO2 | 10 G
SDP

Catalog No. 5000210537
Encompass_Preferred

(S)-2-amino-2-(2-chlorophenyl)acetic acid is the S-enantiomer of 2-amino-2-(2-chlorophenyl)acetic acid, a substituted glycine derivative used as a research reagent in stereoselective synthesis and medicinal chemistry.

  • Chiral amino acid derivative, S-enantiomer.
  • Cas number 141315-50-6.
  • Purity ≥97.0%.
  • Molecular formula C8H8ClNO2; molecular weight 185.61 g·mol⁻¹.
  • Available in 5 g and 10 g packages.
  • Intended for research use only; not for human or clinical use.

Catalog No. 50-002-10537 Supplier Medchemexpress LLC Supplier No. HYW05002310G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(S)-2-amino-2-(2-chlorophenyl)acetic acid is the S-enantiomer of 2-amino-2-(2-chlorophenyl)acetic acid, a substituted glycine derivative used as a research reagent in stereoselective synthesis and medicinal chemistry.

  • Chiral amino acid derivative, S-enantiomer.
  • Cas number 141315-50-6.
  • Purity ≥97.0%.
  • Molecular formula C8H8ClNO2; molecular weight 185.61 g·mol⁻¹.
  • Available in 5 g and 10 g packages.
  • Intended for research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.