Learn More
Medchemexpress LLC (S)-2-amino-2-(thiophen-2-yl)acetic acid | 43189-45-3 | MFCD00078081 | 157.19 | C6H7NO2S | 5 G
SDP
Catalog No. 5000299908
Encompass_Preferred
(S)-2-amino-2-(thiophen-2-yl)acetic acid is the S-enantiomer of 2-thienylglycine, a chiral amino acid derivative used as a research reagent and synthetic building block in medicinal chemistry and peptide synthesis. It is supplied as a dry powder in laboratory-scale quantities for use in synthesis and analytical studies.
  • Chiral S-enantiomer suitable for stereospecific synthesis.
  • Useful building block for small-molecule and peptide synthesis.
  • Compatible with standard peptide coupling and derivatization reactions.
  • Available in multiple laboratory package sizes, including small and mid-scale quantities.
  • Solid form facilitates handling and storage for research workflows.
Catalog No. 50-002-99908 Supplier Medchemexpress LLC Supplier No. HYW0172565G
(S)-2-amino-2-(thiophen-2-yl)acetic acid is the S-enantiomer of 2-thienylglycine, a chiral amino acid derivative used as a research reagent and synthetic building block in medicinal chemistry and peptide synthesis. It is supplied as a dry powder in laboratory-scale quantities for use in synthesis and analytical studies.
  • Chiral S-enantiomer suitable for stereospecific synthesis.
  • Useful building block for small-molecule and peptide synthesis.
  • Compatible with standard peptide coupling and derivatization reactions.
  • Available in multiple laboratory package sizes, including small and mid-scale quantities.
  • Solid form facilitates handling and storage for research workflows.