Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S)-2-amino-2-(thiophen-2-yl)acetic acid | 43189-45-3 | MFCD00078081 | >99.0% | 157.19 g/mol | C6H7NO2S | 1 G
SDP

Catalog No. 5000301185
Encompass_Preferred

(S)-2-Amino-2-(thiophen-2-yl)acetic acid is the S-enantiomer of 2-amino-2-(thiophen-2-yl)acetic acid, a chiral amino acid derivative used as a research reagent and building block in medicinal chemistry and peptide synthesis. Supplied as a solid, it has molecular formula C6H7NO2S and molecular weight 157.19 g/mol. Recommended storage is 4°C, protected from light; in solvent: -80°C (up to 6 months) or -20°C (up to 1 month).

  • Chiral S-enantiomer for stereospecific synthesis.
  • Solid form for convenient handling and measurement.
  • Useful building block for medicinal chemistry and peptide synthesis.
  • Molecular formula C6H7NO2S, molecular weight 157.19 g/mol.
  • Recommended storage at 4°C; protect from light; in solvent: -80°C (6 months) or -20°C (1 month).

Catalog No. 50-003-01185 Supplier Medchemexpress LLC Supplier No. HYW0172561G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(S)-2-Amino-2-(thiophen-2-yl)acetic acid is the S-enantiomer of 2-amino-2-(thiophen-2-yl)acetic acid, a chiral amino acid derivative used as a research reagent and building block in medicinal chemistry and peptide synthesis. Supplied as a solid, it has molecular formula C6H7NO2S and molecular weight 157.19 g/mol. Recommended storage is 4°C, protected from light; in solvent: -80°C (up to 6 months) or -20°C (up to 1 month).

  • Chiral S-enantiomer for stereospecific synthesis.
  • Solid form for convenient handling and measurement.
  • Useful building block for medicinal chemistry and peptide synthesis.
  • Molecular formula C6H7NO2S, molecular weight 157.19 g/mol.
  • Recommended storage at 4°C; protect from light; in solvent: -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.