Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-2-Amino-3-(1-naphthyl)propanoic Acid 99% (99% e.e.), Thermo Scientific™

Catalog No. AC290122500
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC290122500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC290122500 Supplier Thermo Scientific Chemicals Supplier No. 290122500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55516-54-6
Molecular Formula C13H13NO2
Molecular Weight (g/mol) 215.25
MDL Number MFCD00066086
InChI Key OFYAYGJCPXRNBL-LBPRGKRZSA-N
Synonym l-1-naphthylalanine, 3-1-naphthyl-l-alanine, h-1-nal-oh, s-2-amino-3-naphthalen-1-yl propanoic acid, 3-1-naphthyl-l-ala, unii-9g930ul92r, naphthalen-2-yl-3-alanine, 2s-2-amino-3-naphthalen-1-yl propanoic acid, 3-naphth-1-yl-d-alanine, 1-naphthyl-l-alanine
PubChem CID 2724883
IUPAC Name (2S)-2-azaniumyl-3-(naphthalen-1-yl)propanoate
SMILES [NH3+][C@@H](CC1=C2C=CC=CC2=CC=C1)C([O-])=O

Specifications

Color Beige to White
Enantiomeric Excess 98.5% min.
Infrared Spectrum Authentic
Quantity 250 mg
Specific Rotation -16.5° to -18.5° (25°C, 589nm) (c=0.5, CH3COOH)
Formula Weight 215.25
Percent Purity ≥98.5%
Physical Form Powder
Chemical Name or Material (S)-(-)-2-Amino-3-(1-naphthyl)propanoic acid, 99% (99% e.e.)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.