Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S)-2-amino-3-(3-nitrophenyl)propanoic acid | 19883-74-0 | >98.0% | 210.19 g/mol | C9H10N2O4 | 10 G
SDP

Catalog No. 5000210736
Encompass_Preferred

(S)-2-amino-3-(3-nitrophenyl)propanoic acid is the S-enantiomer of 3-nitrophenylalanine, a nitro-substituted phenylalanine derivative used as a building block in research. The material is supplied as a white to off-white solid and is intended for research use only.

  • S-enantiomer of 3-nitrophenylalanine.
  • Phenylalanine derivative used as a chemical building block.
  • Cas number 19883-74-0.
  • Chemical formula C9H10N2O4.
  • Molecular weight 210.19 g/mol.
  • Purity greater than 98.0% (as listed by distributors).
  • Soluble at 10 mM in DMSO (reported solubility).
  • Typical pack sizes include 1 g, 5 g, 10 g, 25 g.

Catalog No. 50-002-10736 Supplier Medchemexpress LLC Supplier No. HYW05049410G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(S)-2-amino-3-(3-nitrophenyl)propanoic acid is the S-enantiomer of 3-nitrophenylalanine, a nitro-substituted phenylalanine derivative used as a building block in research. The material is supplied as a white to off-white solid and is intended for research use only.

  • S-enantiomer of 3-nitrophenylalanine.
  • Phenylalanine derivative used as a chemical building block.
  • Cas number 19883-74-0.
  • Chemical formula C9H10N2O4.
  • Molecular weight 210.19 g/mol.
  • Purity greater than 98.0% (as listed by distributors).
  • Soluble at 10 mM in DMSO (reported solubility).
  • Typical pack sizes include 1 g, 5 g, 10 g, 25 g.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.