Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S)-2-Amino-3-(4-hydroxyphenyl)propionic acid tert-butyl ester | 16874-12-7 | 99.9% | 237.30 | 10 G
SDP

Catalog No. 5000404255
Encompass_Preferred

(S)-2-Amino-3-(4-hydroxyphenyl)propionic acid tert-butyl ester is a tyrosine derivative. Amino acids and amino acid derivatives have been commercially used as ergogenic supplements. They influence the secretion of anabolic hormones, supply of fuel during exercise, mental performance during stress related tasks and prevent exercise induced muscle damage. They are recognized to be beneficial as ergogenic dietary substances. Amino acid derivatives can influence the secretion of anabolic hormones, supply fuel during exercise, improve mental performance during stress-related tasks, and prevent exercise-induced muscle damage.

  • CAS No.: 16874-12-7
  • Appearance: Solid
  • Color: Off-white to light yellow
  • For research use only

Catalog No. 50-004-04255 Supplier Medchemexpress LLC Supplier No. HY3451910G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(S)-2-Amino-3-(4-hydroxyphenyl)propionic acid tert-butyl ester is a tyrosine derivative. Amino acids and amino acid derivatives have been commercially used as ergogenic supplements. They influence the secretion of anabolic hormones, supply of fuel during exercise, mental performance during stress related tasks and prevent exercise induced muscle damage. They are recognized to be beneficial as ergogenic dietary substances. Amino acid derivatives can influence the secretion of anabolic hormones, supply fuel during exercise, improve mental performance during stress-related tasks, and prevent exercise-induced muscle damage.

  • CAS No.: 16874-12-7
  • Appearance: Solid
  • Color: Off-white to light yellow
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.