Learn More
Medchemexpress LLC (S)-2-amino-3-(benzo[b]thiophen-3-yl)propanoic acid | 72120-71-9 | MFCD00079684 | >98.5% | 221.28 g/mol | C11H11NO2S | 1 G
SDP
Catalog No. 5000299603
Encompass_Preferred
(S)-2-Amino-3-(benzo[b]thiophen-3-yl)propanoic acid is the (S)-enantiomer of 3-benzothienylalanine, an alanine derivative bearing a benzothiophene substituent. Supplied as a research-grade amino acid, it is used as a building block in medicinal chemistry and peptide synthesis to introduce a benzothiophene side chain for SAR and conformational studies.
  • Enantiomerically defined (S)-configuration for stereospecific synthesis.
  • Benzothiophene side chain to modulate hydrophobic and aromatic interactions.
  • Suitable as an amino acid building block for peptide and small-molecule synthesis.
  • White to off-white solid, convenient for handling and storage.
  • Provided for research use only.
  • Available in multiple pack sizes for screening and scale-up.
Catalog No. 50-002-99603 Supplier Medchemexpress LLC Supplier No. HYW0227961G
(S)-2-Amino-3-(benzo[b]thiophen-3-yl)propanoic acid is the (S)-enantiomer of 3-benzothienylalanine, an alanine derivative bearing a benzothiophene substituent. Supplied as a research-grade amino acid, it is used as a building block in medicinal chemistry and peptide synthesis to introduce a benzothiophene side chain for SAR and conformational studies.
  • Enantiomerically defined (S)-configuration for stereospecific synthesis.
  • Benzothiophene side chain to modulate hydrophobic and aromatic interactions.
  • Suitable as an amino acid building block for peptide and small-molecule synthesis.
  • White to off-white solid, convenient for handling and storage.
  • Provided for research use only.
  • Available in multiple pack sizes for screening and scale-up.