Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-2-Amino-3-methyl-1,1-diphenyl-1-butanol 98.0+%, TCI America™
SDP

Catalog No. A25045G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A25045G 5 g
A25041G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A25045G Supplier TCI America Supplier No. A25045G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 78603-95-9
Molecular Formula C17H21NO
Molecular Weight (g/mol) 255.36
MDL Number MFCD01318248
InChI Key LNQVZZGGOZBOQS-UHFFFAOYNA-N
PubChem CID 854145
IUPAC Name 2-amino-3-methyl-1,1-diphenylbutan-1-ol
SMILES CC(C)C(N)C(O)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 98°C
Color White-Yellow
Quantity 5 g
Formula Weight 255.36
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-(-)-2-Amino-3-methyl-1,1-diphenyl-1-butanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.