Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S)-2-amino-4-(4-((3,4-dichlorobenzyl)oxy)phenyl)-2-methylbutan-1-ol | 2748196-63-4 | MFCD00006370 | 97.0% | 354.27 | C18H21Cl2NO2 | 100MG
SDP

Catalog No. 5000237847
Encompass_Preferred

P053 is a research-grade small molecule inhibitor of ceramide synthase 1 (CerS1) used to study lipid metabolism and mitochondrial fatty acid oxidation. It is a potent, non-competitive, and selective CerS1 inhibitor (IC50 0.5 μM) supplied as a purified solid for laboratory research and intended for research use only.

  • Selective CerS1 inhibitor with reported IC50 of 0.5 μM.
  • Modulates mitochondrial fatty acid oxidation and whole-body adiposity regulation.
  • Chemical formula C18H21Cl2NO2, molecular weight 354.27.
  • Purity 97.0% and solid, white to off-white appearance.
  • Soluble in DMSO at 100 mg/mL (ultrasonic and warming; heat to 60°C).
  • Storage: powder -20°C for 3 years, 4°C for 2 years; in solvent -80°C for 6 months, -20°C for 1 month.
  • For research use only; not for clinical or human use.

Catalog No. 50-002-37847 Supplier Medchemexpress LLC Supplier No. HY126015100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

P053 is a research-grade small molecule inhibitor of ceramide synthase 1 (CerS1) used to study lipid metabolism and mitochondrial fatty acid oxidation. It is a potent, non-competitive, and selective CerS1 inhibitor (IC50 0.5 μM) supplied as a purified solid for laboratory research and intended for research use only.

  • Selective CerS1 inhibitor with reported IC50 of 0.5 μM.
  • Modulates mitochondrial fatty acid oxidation and whole-body adiposity regulation.
  • Chemical formula C18H21Cl2NO2, molecular weight 354.27.
  • Purity 97.0% and solid, white to off-white appearance.
  • Soluble in DMSO at 100 mg/mL (ultrasonic and warming; heat to 60°C).
  • Storage: powder -20°C for 3 years, 4°C for 2 years; in solvent -80°C for 6 months, -20°C for 1 month.
  • For research use only; not for clinical or human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.