Learn More
Medchemexpress LLC (S)-2-amino-4-(4-((3,4-dichlorobenzyl)oxy)phenyl)-2-methylbutan-1-ol | 2748196-63-4 | MFCD00006370 | 97.0% | 354.27 | C18H21Cl2NO2 | 50MG
SDP
Catalog No. 5000239147
Encompass_Preferred
This compound is a selective, non-competitive inhibitor of ceramide synthase 1 (CerS1) used as a research tool to study lipid metabolism and mitochondrial fatty acid oxidation. It displays an IC50 of approximately 0.5 μM against CerS1 and has been applied in cellular and in vivo models investigating muscle fatty acid oxidation and whole-body adiposity regulation.
  • Selective CerS1 inhibition with reported IC50 ~0.5 μM.
  • Non-competitive mechanism suitable for target validation studies.
  • Appropriate for in vitro and in vivo research applications.
  • Chemical formula C18H21Cl2NO2 and molecular weight 354.27.
  • Typical purity 97.0% as supplied.
  • Offered as solid quantities and as a DMSO solution for dosing flexibility.
Catalog No. 50-002-39147 Supplier Medchemexpress LLC Supplier No. HY12601550MG
This compound is a selective, non-competitive inhibitor of ceramide synthase 1 (CerS1) used as a research tool to study lipid metabolism and mitochondrial fatty acid oxidation. It displays an IC50 of approximately 0.5 μM against CerS1 and has been applied in cellular and in vivo models investigating muscle fatty acid oxidation and whole-body adiposity regulation.
  • Selective CerS1 inhibition with reported IC50 ~0.5 μM.
  • Non-competitive mechanism suitable for target validation studies.
  • Appropriate for in vitro and in vivo research applications.
  • Chemical formula C18H21Cl2NO2 and molecular weight 354.27.
  • Typical purity 97.0% as supplied.
  • Offered as solid quantities and as a DMSO solution for dosing flexibility.